missing translation for 'onlineSavingsMsg'
Learn More

Contract Required Detection Limits (CRDL) Standard, Specpure™, Thermo Scientific Chemicals

294.47 € - 625.00 €

Specifications

Chemical Name or Material Contract Required Detection Limits (CRDL) Standard
Concentration Matrix: 5% HNO3/tr. tartaric acid
MDL Number MFCD00146530
UN Number UN3264
Grade Specpure™
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313627
View Documents
Thermo Scientific Alfa Aesar
035420.AE
100 mL
294.47 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11425010
View Documents
Thermo Scientific Alfa Aesar
035420.AP
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Contract Required Detection Limits (CRDL) Standard
MFCD00146530
Specpure™
Matrix: 5% HNO3/tr. tartaric acid
UN3264
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.