Learn More
Buerkle™ Close-it™ Control Seal Food & Pharma
Control Seal Close-it Food and Pharma with an adhesive approved for use with foodstuffs. Reliably prevents products from leaking out after sampling.
Brand:  Buerkle™ 5303-2023
65.08 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15356287
  • 82.45 € / Pack of 500
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reliably prevents products from leaking out after sampling.
  • The adhesive complies with the European guidelines for foodstuffs.
  • Aluminum foil barrier layer
  • Suitable for sampling foodstuffs, pharmaceuticals, cosmetics, food additives, even frozen goods – in short, for all applications in which cleanliness is particularly important.
  • According to FDA Guidelines
  • Particularly suited for the food and pharma industry
Specifications
Specifications
FDA
Control Seal
Square
0.8965 kg
Black
Foodstuffs
95 x 95 mm