Learn More
Epredia™ Cool-Cut with universal cassette clamp
Maintain a cool, consistent temperature for paraffin blocks during the sectioning process with the Epredia™ Cool-Cut accessory.
Brand: Epredia™ 771120
3028.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Directly fits onto the specimen cylinder of the rotary microtome line
- Ability to be equipped as a universal cassette clamp or standard specimen clamp
- Effective when extended cutting periods on the microtome are required to “step section” or “serial section” through the paraffin block
Includes:
- Peltier device with integrated specimen clamp
- Counter-balance weights
- Power supply
Specifications
With standard clamp | |
Microtome Accessory | |
Epredia Microtomes |