missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolBox™ XT Systems
BIOPHARMA

Product Code. p-6827093
Click to view available options
Color:
Green
Orange
Pink
Purple
Includes:
CoolBox 2XT System, CoolRack PCR96, SBS-Compatible CoolRack XT M24
CoolBox XT System, SBS-Compatible CoolRack XT M-PCR
CoolBox XT System, SBS-Compatible CoolRack XT M24
Holder and One Cooling Core
Holder and Two Cooling Cores
Product Line:
CoolBox 2XT
CoolBox 2XT PCR system
CoolBox XT
CoolBox XT M24 system
CoolBox XT PCR Strip system
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 15562791

missing translation for 'mfr': Corning™ 432021

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provide sample cooling or freezing without ice, electricity, or batteries.

Corning CoolBox XT systems are bench top cooling workstations that provide sample cooling or freezing without ice, electricity or batteries. CoolBox systems are versatile and accommodate a variety of sample formats and temperatures. The modular design enables the use of CoolRack™ and CoolSink™ thermo-conductive sample modules to hold microfuge tubes, cryogenic vials, PCR tubes or plates, assay plates and more.

  • Consists of a base, cooling cartridge/core and a lid
  • Includes a magnetized collar and a 1° to 8°C temperature indicator for visual assurance of proper temperature
  • To use with any 30 place Corning CoolRack Tubes Module
  • The internal cooling or freezing cores provide 0.5° to 4°C for over 10 hours with lid open and over 16 hours with lid closed
  • CoolBox systems may also be used with dry ice to create a compact, portable, snap freezing workstation for bacteria, virus or proteins.
TRUSTED_SUSTAINABILITY

Specifications

Description Benchtop cooling system
Color Purple
Type Cool Box
For Use With (Application) Enables Cooling for 16 Hours, Freezing for 8 Hours
Holds 30 Place
Includes Holder and One Cooling Core
Product Line CoolBox XT
Weight (Metric) 3 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt