Learn More
Corning™ CoolCell™ Cell Freezing Vial Containers

Controls rate of freezing to -1°C/minute when placed in a -80°C freezer.
Brand: Corning™ 432000
149.43 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing

Specifications
High cell recovery, cell viability | |
Foam | |
Freezer Box | |
1/Pk., 1/Cs. |
Purple | |
Round | |
12 (Standard 1 to 2 mL Cryogenic Vials) | |
0.19 kg |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.