Learn More
Corning™ CoolCell™ 2mL Filler Vials for CoolCell™ LX and FTS30 Containers
Suitable for repeated use. Corning™ CoolCell™ 2mL Filler Vials ensure cell freezing rate consistency and uniform results when using CoolCell containers.
Brand: Corning™ 432076
27.69 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Corning CoolCell, in combination with a -80°C freezer, will provide alcohol-free freezing at the rate of -1°C/minute that is ideal for cryopreservation of most cells and cell lines. Using a combination of uniform-density cross-linked polyethylene foam, a solid state core, and radial vial symmetry, freezing profiles are consistent and reproducible. The foam is non-absorbent and will impose negligible change in the freezer environment; thereby protecting nearby frozen samples. The removal of vials is accomplished by tipping the device upside down so the vials slide out.
Features: Unique features of controlled-rate freezing with Corning™ CoolCell™ include:
- Ease of use
- Alcohol and fluid-free freezing
- Lower cost of use than alcohol-based devices
- High cell recovery and cell viability
- Reproducibility
- Simple, consistent way to standardize controlled-rate freezing
Specifications
White | |
2 mL | |
6 | |
CoolCell LX, CoolCell FTS30 5ml LX Containers |
Clear | |
Glycerol | |
6 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.