Learn More
Corning™ Extension Collar for CoolBox™ 2XT System
BIOPHARMA
For use with CoolBox™ 2XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
Brand:  Corning™ 432087
 View more versions of this product
Product Code. 15522861
  • 132.00 € / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Alert:
Lid closure not possible with addition of extension collar
Description
Description
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Extension Collar
Purple
1/Cs.
CoolBox 2XT System
26.5 x 20.0 x 7.0 cm
Magnetized Extension Collar
1/Pk.
To extend height of CoolBox™ ice-free cooling systems
21.0 x 14.3 x 7.0 cm