missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cooling Tube for Electrophoresis Units

For use circulating coolant via the MultiTemp III Thermostatic Circulator to electrophoresis units such as the Multiphor II

Brand:  Cytiva 80110656

246.05 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


Product Code. 11882190

  • 267.00 € / 4m
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cooling tubing: 8mm i.d.,12 mm o.d. in 4 m pack size

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MultiTemp III Thermostatic Circulator Electrophoresis Unit
Cooling Tube
157.48 in.
400 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.