missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ Tall Modules
BIOPHARMA

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Brand:  Corning™ 432060

311.04 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15552831

  • 324.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.

  • Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
  • All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
  • SBS-compatible
TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 x 5 mL centrifuge tubes, SBS-compatible
CoolRack XT 5 mL
Tall Module
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number04216027 Certificate TypeCertificate of Quality Date31/12/2024 Catalog Number432060
Lot Number04216027 Certificate TypeCertificate of Compliance Date30/11/2022 Catalog Number432060
Lot Number04216027 Certificate TypeCertificate of Analysis Date12/05/2022 Catalog Number432060
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ CoolRack™ Tall Modules > CoolRack XT 5 mL; 12 x 5 mL centrifuge tubes, SBS-compatible

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.