Learn More
Corning™ CoolRack™ Tall Modules

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
Brand: Corning™ 432061
312.91 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
Corning CoolRack thermo-conductive centrifuge tube modules eliminate variability which originates from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources. Place the CoolRack module directly onto a temperature source between -196°C to >100°C and it will rapidly adapt to that temperature.
- Ensures ± 0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.
- All CoolRack modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
- Suggested applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen.
- SBS-compatible

Specifications
9 x 15 mL Centrifuge Tubes | |
CoolRack 15 mL | |
Tall Module |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.