missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coomassie Brilliant Blue R-250, Fisher BioReagents
26.30 € - 83.85 €
Chemical Identifiers
CAS | 6104-59-2 |
---|---|
Molecular Formula | C45H44N3NaO7S2 |
Molecular Weight (g/mol) | 825.97 |
MDL Number | MFCD00041762 |
InChI Key | HAEGGLCXLFWTBE-UHFFFAOYSA-M |
Synonym | Coomassie Blue R-250 |
PubChem CID | 61365 |
IUPAC Name | sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
SMILES | [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 |
Description
Brilliant Blue R-250 is commonly used for staining proteins in polyacrylamide gels.Chemical Identifiers
6104-59-2 | |
825.97 | |
HAEGGLCXLFWTBE-UHFFFAOYSA-M | |
61365 | |
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 |
C45H44N3NaO7S2 | |
MFCD00041762 | |
Coomassie Blue R-250 | |
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
Specifications
174°C | |
700g(-1) cm(-1) min. | |
MFCD00041762 | |
Pass Test | |
[Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1 | |
825.97 | |
590 to 594nm | |
5% max. | |
42660 | |
6.2 | |
Brilliant Blue R-250 |
6104-59-2 | |
C45H44N3NaO7S2 | |
Coomassie Blue R-250 | |
HAEGGLCXLFWTBE-UHFFFAOYSA-M | |
sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate | |
61365 | |
825.96 | |
0.25g/cm³ | |
Brown/Red | |
Powder |
Safety and Handling
Recommended Storage : RT
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title