Learn More
Copper based low temperature water gas shift catalyst, HiFUEL™ W220, Thermo Scientific™
77.00 € - 307.00 €
Specifications
Chemical Name or Material | Copper based low temperature water gas shift catalyst, HiFUEL W220 |
---|---|
Color | Black |
Appearance | Black pellets |
Form | Pellet |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Copper based low temperature water gas shift catalyst, HiFUEL W220 | |
Black | |
Black pellets | |
Pellet |
Safety and Handling
H400-H410; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.