missing translation for 'onlineSavingsMsg'
Learn More
Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals Product Code.: 15402887

Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15402887
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15402887

Brand: Thermo Scientific Alfa Aesar 047162.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291300-50-0
Molecular Formula C2CuF6NO4S2
Molecular Weight (g/mol) 343.681
MDL Number MFCD23380170
InChI Key MSFHCZUIBARZOS-UHFFFAOYSA-N
Synonym copper trifluoromethanesulfonimide
PubChem CID 134159300
IUPAC Name bis(trifluoromethylsulfonyl)azanide;copper(1+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+]

Specifications

Color Blue
Linear Formula [(CF3SO2)2N]Cu
Quantity 1 g
UN Number UN3261
Sensitivity Very Hygroscopic
Formula Weight 343.68
Physical Form Liquid or Solid
Chemical Name or Material Copper bis(trifluoromethylsulfonyl)imide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN08J006 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number047162.03,047162.06,047162.MD
Lot NumberR16I011 Certificate TypeCertificate of Analysis Date13/06/2022 Catalog Number047162.03,047162.06,047162.MD
Lot NumberP05A043 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number047162.03,047162.06,047162.MD
Lot NumberX27D082 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number047162.03,047162.06,047162.MD
compliance-icons
Product Identifier
  • Copper bis(trifluoromethylsulfonyl)imide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Copper bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.