missing translation for 'onlineSavingsMsg'
Learn More

Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals

Product Code. 15422887
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 291300-50-0
Molecular Formula C2CuF6NO4S2
Molecular Weight (g/mol) 343.681
MDL Number MFCD23380170
InChI Key MSFHCZUIBARZOS-UHFFFAOYSA-N
Synonym copper trifluoromethanesulfonimide
PubChem CID 134159300
IUPAC Name bis(trifluoromethylsulfonyl)azanide;copper(1+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Cu+]

Specifications

Color Blue
Linear Formula [(CF3SO2)2N]Cu
Quantity 250 mg
UN Number UN3261
Sensitivity Very Hygroscopic
Formula Weight 343.68
Physical Form Liquid or Solid
Chemical Name or Material Copper bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.