missing translation for 'onlineSavingsMsg'
Learn More
Copper foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11356708

Copper foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11356708
20 x 100 cm
Click to view available options
Quantity:
20 x 20 cm
20 x 100 cm
This item is not returnable. View return policy

Product Code. 11356708

Brand: Thermo Scientific Alfa Aesar 013380.CV

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Assay Percent Range 99.9% (metals basis)
Quantity 20 x 100 cm
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Copper foil (Annealed), 0.127mm (0.005 in.) thick, Annealed, Temper: soft

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT20K010 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog Number013380.CV,013380.CU
Lot NumberZ26K019 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number013380.CV,013380.CU
Lot NumberY15K038 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog Number013380.CV,013380.CU
Lot NumberX10K045 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number013380.CV,013380.CU
Lot NumberU30K019 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number013380.CV,013380.CU

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper foil, 0.127mm (0.005in) thick, annealed, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.