missing translation for 'onlineSavingsMsg'
Learn More
Copper foil, 99.999% (metals basis), Thermo Scientific Chemicals, Puratronic™ Product Code.: 11381498

Copper foil, 99.999% (metals basis), Thermo Scientific Chemicals, Puratronic™

Product Code. 11381498
25 x 200 mm
Click to view available options
Quantity:
25 x 200 mm
25 x 1000 mm
This item is not returnable. View return policy

Product Code. 11381498

Brand: Thermo Scientific Alfa Aesar 010950.HV

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: approx. 1.1g/25x200mm

Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs and heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clay. It is also used as catalyst in hydrodesulfurization and in Ziegler-Natta polymerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored foil
Assay Percent Range 99.999% (Metals basis)
Quantity 25 x 200 mm
Molecular Weight (g/mol) 63.55
Odor Odorless
Vapor Pressure 0 hPa
Form Foil
Chemical Name or Material Copper

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN29K009 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number010950.HV
Lot NumberM22G029 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number010950,010950.HV
Lot NumberT28I010 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog Number010950.HV
Lot NumberM22G028 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010950.HV
Lot NumberA10X013 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010950.HV

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper foil, 99.999% (metals basis), Thermo Scientific Chemicals, Puratronic™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.