Learn More
Copper(I) bromide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
95.58 € - 315.57 €
Chemical Identifiers
CAS | 7787-70-4 |
---|---|
Molecular Formula | BrCu |
Molecular Weight (g/mol) | 143.45 |
MDL Number | MFCD00010969 |
InChI Key | NKNDPYCGAZPOFS-UHFFFAOYSA-M |
Synonym | copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide |
PubChem CID | 24593 |
IUPAC Name | bromocopper |
SMILES | [Cu+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328678
|
Thermo Scientific Alfa Aesar
040752.03 |
1 g |
95.58 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338678
|
Thermo Scientific Alfa Aesar
040752.06 |
5 g |
315.57 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as effective catalyst for tetrahydropyranylation of alcohols. It can also be used in combination with palladium in catalytic synthesis of 3-haloindoles through annulation process. It is widely used in the synthesis of organic fine chemicals. The compound is widely used in the synthesis of organic compounds. It is also useful for copper-mediated coupling reactions. A silica gel support copper bromide hexa methyltriethylenetetramine complex has shown useful application for the atom transfer radical polymerization of methyl methacrylate in toluene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7787-70-4 | |
143.45 | |
NKNDPYCGAZPOFS-UHFFFAOYSA-M | |
24593 | |
[Cu+].[Br-] |
BrCu | |
MFCD00010969 | |
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide | |
bromocopper |
Specifications
1345°C | |
7787-70-4 | |
Powder | |
BrCu | |
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide | |
NKNDPYCGAZPOFS-UHFFFAOYSA-M | |
bromocopper | |
24593 | |
Air Sensitive | |
4.98 g/cm3 |
492°C | |
Green | |
99.998% (Metal basis) | |
MFCD00010969 | |
Soluble in hydrogen bromide,hydrochloric acid,and ammonium hydroxide. Slightly soluble in water. Insoluble in acetone. | |
[Cu+].[Br-] | |
143.45 | |
143.45 | |
Light green powder | |
Copper(I) bromide |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'dotInformation' : DOT Class: 9, Packing Group: III
missing translation for 'einecsNumber' : 232-131-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Copper(I) bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : copper bromide
RUO – Research Use Only