missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Chloride, Honeywell™
Reagent Grade, 97%
35.87 € - 271.77 €
Chemical Identifiers
CAS | 7758-89-6 |
---|---|
Molecular Formula | ClCu |
Molecular Weight (g/mol) | 98.996 |
MDL Number | MFCD00010971 |
InChI Key | OXBLHERUFWYNTN-UHFFFAOYSA-M |
Synonym | cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech |
PubChem CID | 62652 |
ChEBI | CHEBI:53472 |
IUPAC Name | chlorocopper |
SMILES | Cl[Cu] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15641950
|
Honeywell Research Chemicals
212946-25G |
25 g | N/A | ||||||
15661950
|
Honeywell Research Chemicals
212946-500G |
500 g | N/A | ||||||
15651950
|
Honeywell Research Chemicals
212946-2KG |
2 kg |
271.77 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7758-89-6 | |
98.996 | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
62652 | |
chlorocopper |
ClCu | |
MFCD00010971 | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
CHEBI:53472 | |
Cl[Cu] |
Specifications
7758-89-6 | |
CuCl | |
2802 | |
Partially soluble in dimethyl sulfide, Insoluble in water | |
Cl[Cu] | |
98.996 | |
CHEBI:53472 | |
97% | |
1.3mmHg (546°C) |
ClCu | |
MFCD00010971 | |
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech | |
OXBLHERUFWYNTN-UHFFFAOYSA-M | |
chlorocopper | |
62652 | |
99g/mol | |
Reagent | |
Copper(I) Chloride |
Safety and Handling
P273-P280-P305 + P351 + P338-P501
H302-H315-H318-H410
missing translation for 'einecsNumber' : 231-842-9
missing translation for 'rtecsNumber' : GL6990000