missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) iodide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent

Specifications
Specifications
Boiling Point | 1290.0°C |
Melting Point | 605.0°C |
Color | Brown-Gray to White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 97.5% min. (Complexometry) |
Linear Formula | CuI |
Fieser | 01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88 |
Merck Index | 15, 2652 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0434829 | Certificate of Analysis | 11/10/2024 | 194900050,194902500,194900010 |
A0466459 | Certificate of Analysis | 26/09/2024 | 194900050,194902500,194900010 |
A0461138 | Certificate of Analysis | 31/03/2024 | 194900050,194902500,194900010 |
A0443880 | Certificate of Analysis | 08/01/2024 | 194900050,194902500,194900010 |
A0423730 | Certificate of Analysis | 03/11/2023 | 194900050,194902500,194900010 |
Safety and Handling


Product Identifier
- Copper(I) iodide
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Copper(I) iodide, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction