missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) iodide, 98%, Thermo Scientific Chemicals

Product Code. 11317357
Click to view available options
Quantity:
250 g
1 kg

It is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-65-4
MDL Number MFCD00010978
Synonym cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Specifications

Boiling Point ∼1290°C
Melting Point 605°C
Color Brown
Physical Form Powder
Quantity 250 g
UN Number UN3077
Merck Index 14,2662
Solubility Information Copper(I) iodide is soluble in ammonia,thiosulfate,sodium and potassium iodide solutions; Insoluble in water and dilute acids.
Formula Weight 190.45
Percent Purity 98%
Odor Weak
Refractive Index 2.346
Sensitivity Air, light, and moisture sensitive
Density 5.63 g/mL
Chemical Name or Material Copper(I) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.