missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) phenylacetylide, Thermo Scientific Chemicals

Product Code. 11303377
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
2 g
missing translation for 'unitSize'
2g
500mg
This item is not returnable. View return policy

Product Code. 11303377

missing translation for 'mfr': Thermo Scientific Alfa Aesar 017538.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Copper(I) phenylacetylide participates in efficient coupling with a range of aryl iodides and bromides to give substituted tolanes which, if possessing an ortho amino or hydroxy group, can be thermolyzed to yield 2-phenylbenzofurans, -indoles, and -furo[3,2-b]pyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13146-23-1
Molecular Formula C8H5Cu
Molecular Weight (g/mol) 164.674
MDL Number MFCD00015578
InChI Key SFLVZUJLUPAJNE-UHFFFAOYSA-N
Synonym copper 1+ ; ethynylbenzene, copper i phenylacetylide, copper i phenylacetylide, min
PubChem CID 6101206
IUPAC Name copper(1+);ethynylbenzene
SMILES [C-]#CC1=CC=CC=C1.[Cu+]

Specifications

Color Yellow
Quantity 2 g
UN Number UN3088
Sensitivity Air and moisture sensitive
Solubility Information Soluble in acetone and acetonitrile; Insoluble in water
Formula Weight 153.68
Physical Form Powder
Chemical Name or Material Copper(I) phenylacetylide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.