missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) carbonate basic, pure, 54-56% Cu

Product Code. 10452391 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10452391

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12069-69-1 | CCuO3·CuO2H2 | 221.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-69-1
Molecular Formula CCuO3·CuO2H2
Molecular Weight (g/mol) 221.1
MDL Number MFCD00010976
Synonym Copper(II)hydroxide carbonate

Specifications

Melting Point 200.0°C
Color Green
Physical Form Fine Crystalline Powder
Quantity 500 g
Linear Formula CuCO3·Cu(OH)2
Fieser 01,163
Merck Index 15, 2621
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acids, ammonia, practically insoluble in alcohol
Formula Weight 221.1
Percent Purity 54 to 56% Cu
Grade Pure
Packaging Glass bottle
Density 4
Chemical Name or Material Copper(II) carbonate basic
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) carbonate basic
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.