missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) chloride dihydrate, 99%, Thermo Scientific Chemicals Product Code.: 11453077

Copper(II) chloride dihydrate, 99%, Thermo Scientific Chemicals

Product Code. 11453077
250 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11453077

Brand: Thermo Scientific Alfa Aesar A16064.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) chloride is used as a catalyst for organic and inorganic reactions, mordant for dyeing and printing materials, pigment for glass, ceramics, as wood preservative, as disinfectant, and as a catalyst in the output of chlorine from hydrogen chloride. It is likewise employed in the oil industry as a purifying agent, in the manufacture of laundry marking inks, in metallurgy to recover mercury from ores, in refining copper, silver and gold, in tinting baths of iron and tin, in photography, in pyrotechnics, and to get rid of lead compounds from gasoline and oils.

Solubility
Soluble in water, methanol and ethanol.

Notes
Incompatible with materials such as oxidizing agents, metals and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10125-13-0
Molecular Formula Cl2CuH4O2
Molecular Weight (g/mol) 170.48
MDL Number MFCD00149674
InChI Key MPTQRFCYZCXJFQ-UHFFFAOYSA-L
Synonym coppertrace, copper ii chloride dihydrate, copper chloride dihydrate, caswell no. 235a, copper 2+ chloride dihydrate, cupric chloride usp, copper chloride cucl2 , dihydrate, epa pesticide chemical code 023701
PubChem CID 61482
ChEBI CHEBI:86318
IUPAC Name dichlorocopper;dihydrate
SMILES O.O.[Cl-].[Cl-].[Cu++]

Specifications

Melting Point 100°C (decomposition)
pH 3.0 to 3.8
Quantity 250 g
UN Number UN2802
Merck Index 14,2633
Solubility Information Soluble in water,methanol and ethanol.
Formula Weight 170.48 (134.45 Anhydrous)
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.51
Chemical Name or Material Copper(II) chloride dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250385 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberA16064.30,A16064.0B
Lot Number10247344 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog NumberA16064.30,A16064.0B,A16064
Lot Number10242986 Certificate TypeCertificate of Analysis Date19/08/2022 Catalog NumberA16064.30,A16064.0B
Lot Number10241988 Certificate TypeCertificate of Analysis Date12/07/2022 Catalog NumberA16064.30,A16064.0B
Lot Number10238210 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA16064.30,A16064.0B
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Copper(2+) chloridedihydrate, Copper chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) chloride dihydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.