missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) ethoxide, Thermo Scientific™ Product Code.: 11313507

Copper(II) ethoxide, Thermo Scientific™

Product Code. 11313507
0.5 g
Click to view available options
Quantity:
0.5 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11313507

Brand: Thermo Scientific Alfa Aesar 018862.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2850-65-9
Molecular Formula C4H10CuO2
Molecular Weight (g/mol) 153.668
MDL Number MFCD00015650
InChI Key CRCKGIUJMFFISH-UHFFFAOYSA-N
Synonym copper ii ethoxide, copper diethoxide, ethanol, copper 2 salt, copper ethoxide, cupric ethoxide, acmc-20aldp, copper 2+ bis ethoxide, copper 2+ ion bis ethoxide
PubChem CID 14419876
IUPAC Name copper;ethanolate
SMILES CC[O-].CC[O-].[Cu+2]

Specifications

Melting Point ∼120°C (decomposition)
Color Green
Odor Alcohol-like
Linear Formula Cu(OC2H5)2
Quantity 0.5 g
UN Number UN3181
Sensitivity Moisture sensitive
Solubility Information Reacts with water and decomposes.
Formula Weight 153.67
Physical Form Powder
Chemical Name or Material Copper(II) ethoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) ethoxide, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.