missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) fluoride, 98%, anhydrous, Thermo Scientific Chemicals

Product Code. 11934171
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-19-7
Molecular Formula CuF2
Molecular Weight (g/mol) 101.54
Synonym Cupric fluoride

Specifications

Melting Point 950.0°C
Color Blue-Gray to Gray
Physical Form Powder
Quantity 100 g
Assay Percent Range 97.5% min
Linear Formula CuF2
Merck Index 15,2628
Solubility Information Solubility in water: 47g/L (20°C). Other solubilities: hydrolyzes in hot water (forming oxyfluoride), soluble in acids and alcohol
Formula Weight 101.54
Percent Purity 98%
Loss on Drying 0.5% max.
Packaging Plastic bottle
Specific Gravity 4.23
Density 4.2300g/mL
Chemical Name or Material Copper(II) fluoride
Show More Show Less
compliance-icons
Product Identifier
  • Copper(II) fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.