missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) isopropoxide, 98% (metals basis), Thermo Scientific Chemicals Product Code.: 11328727

Copper(II) isopropoxide, 98% (metals basis), Thermo Scientific Chemicals

Product Code. 11328727
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11328727

Brand: Thermo Scientific Alfa Aesar 035821.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23578-23-6
Molecular Formula C6H14CuO2
Molecular Weight (g/mol) 181.72
MDL Number MFCD00074982
InChI Key VNGORJHUDAPOQZ-UHFFFAOYSA-N
Synonym copper ii isopropoxide, acmc-20aldx, copper ii diisopropoxide, copper 2+ dipropan-2-olate, 2-propanol,copper 2+ salt 9ci, copper 2+ bis propan-2-olate, copper 2+ ion bis propan-2-olate
PubChem CID 14389592
IUPAC Name copper;propan-2-olate
SMILES [Cu++].CC(C)[O-].CC(C)[O-]

Specifications

Quantity 1 g
UN Number UN3180
Sensitivity Moisture Sensitive
Formula Weight 181.73
Percent Purity 98%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Copper(II) isopropoxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) isopropoxide, 98% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.