missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) methacrylate, tech., Thermo Scientific Chemicals

Product Code. 11358758
Click to view available options
Quantity:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 19662-59-0
Molecular Formula C4H5CuO2
Molecular Weight (g/mol) 148.63
MDL Number MFCD00080547,MFCD00156451,MFCD00080547
InChI Key BFHDHRZKYIRNRP-UHFFFAOYSA-M
PubChem CID 121233728
IUPAC Name copper;2-methylprop-2-enoic acid;hydrate
SMILES [Cu++].CC(=C)C([O-])=O

Spécification

Color Blue-Green
Linear Formula [H2C=C(CH3)CO2]2Cu
Quantity 5 g
Formula Weight 233.71
Grade Technical
Physical Form Powder
Chemical Name or Material Copper(II) methacrylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'