missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) Nitrate Trihydrate, 95+%, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Melting Point | 114°C |
Color | Blue |
pH | 4 |
Physical Form | Solid |
Quantity | 250 g |
Formula Weight | 241.6g/mol |
Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Copper(II) nitrate, trihydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H272-May intensify fire; oxidiser.
- H314-Causes severe skin burns and eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Copper(II) nitrate,trihydrate (1:2:3),Cupric nitrate
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'