missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) nitrate trihydrate, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 10592031
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-43-3
MDL Number MFCD00149671
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratrem
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]

Specifications

Color Brown to Cream
Physical Form Fine Powder
Quantity 5 g
Assay Percent Range 99%
Molecular Formula CuH6N2O9
Linear Formula Cu(NO3)2·3H2O
Fieser 17,366
Merck Index 15, 2633
Molecular Weight (g/mol) 241.599
Formula Weight 241.6
Percent Purity 98.5 to 104.0% (Iodometry)
Grade Analytical
Packaging Glass Bottle
Chemical Name or Material Copper(II) nitrate trihydrate, p.a.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.