missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) nitrate trihydrate, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 114.0°C |
Quantity | 1 kg |
Assay Percent Range | 98.5 to 104% (Iodometry) |
Linear Formula | Cu(NO3)2·3H2O |
Merck Index | 15,2633 |
Solubility Information | Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate |
Formula Weight | 241.6 |
Percent Purity | 99% |
Grade | Pure |
Chemical Name or Material | Copper(II) nitrate trihydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0471702 | Certificate of Analysis | 10/04/2025 | 453960010,453960050 |
A0466245 | Certificate of Analysis | 01/10/2024 | 453960010,453960050 |
A0461960 | Certificate of Analysis | 24/04/2024 | 453960010,453960050 |
A0450984 | Certificate of Analysis | 16/04/2024 | 453960010,453960050 |
A0447983 | Certificate of Analysis | 16/04/2024 | 453960010,453960050 |
Safety and Handling



Product Identifier
- Copper(II) nitrate, trihydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Copper(II) nitrate trihydrate, 99%, pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction