missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxalate hemihydrate, 98%, Thermo Scientific Chemicals

Product Code. 11428306
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II) oxalate hemihydrate is used as a catalyst for organic reactions; as stabilizer for acetylated polyformaldehyde; in anticaries compositions; in seed treatments to repel birds and rodents.

Solubility
Soluble in NH{4}OH

Notes
Store in cool, dry, and well-ventilated area. Store away from oxidizing agents. Incompatible with acitve metals.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5893-66-3
Molecular Formula C4H8Cu2O10
Molecular Weight (g/mol) 343.19
MDL Number MFCD00150451
InChI Key NNZNHNMQOYUANB-UHFFFAOYSA-N
Synonym cupric oxalate, copper oxalate hemihydrate
PubChem CID 122130000
IUPAC Name copper;oxalic acid;hydrate
SMILES O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O

Spécification

Assay Percent Range 98%
Quantity 250 g
UN Number UN3288
Merck Index 14,2645
Solubility Information Soluble in NH4OH
Formula Weight 160.57 (151.51 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Copper(II) oxalate hemihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'