missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) oxide, ACS reagent, powder, Thermo Scientific Chemicals

31.36 € - 311.55 €

Chemical Identifiers

CAS 1317-38-0
Molecular Formula CuO
Molecular Weight (g/mol) 79.55
MDL Number MFCD00010979
InChI Key KKCXRELNMOYFLS-UHFFFAOYSA-N
Synonym cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
PubChem CID 14829
SMILES [O--].[Cu++]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10528702
View Documents
Thermo Scientific Acros
405860250
25 g Glass Bottle
31.36 €
25g
Please to purchase this item. Need a web account? Register with us today!
10137510
View Documents
Thermo Scientific Acros
405862500
250 g Plastic bottle
81.61 €
250g
Please to purchase this item. Need a web account? Register with us today!
10288803
View Documents
Thermo Scientific Acros
405860010
1 kg Plastic bottle
311.55 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1317-38-0
79.55
KKCXRELNMOYFLS-UHFFFAOYSA-N
14829
CuO
MFCD00010979
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
[O--].[Cu++]

Specifications

1326.0°C
Black
CuO
MFCD00010979
15, 2636
Solubility in water: insoluble. Other solubilities: soluble in dil.acids - slowly soluble in nh3, soluble in alkali cyanides and (nh4)2co3 solutions, practically insoluble in alcohol
[O--].[Cu++]
79.55
79.54
ACS Reagent
1317-38-0
Powder
CuO
04,108; 05,161
cupric oxide, copper ii oxide, copper oxide, copper monoxide, banacobru ol, chrome brown, copper brown, copper monooxide, black copper oxide
KKCXRELNMOYFLS-UHFFFAOYSA-N
oxocopper
14829
≥99.0%
Copper(II) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 215-269-1

missing translation for 'rtecsNumber' : GL7900000

missing translation for 'tsca' : TSCA

Product Identifier
  • Copper(II) oxide

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.