missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, 98+%, for analysis, anhydrous

Product Code. 10174260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10174260

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-98-7 | CuO4S | 159.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.6
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Melting Point 200.0°C
Color Gray to Green
Physical Form Fine Powder
Quantity 100 g
Assay Percent Range 98% min. (Iodometry)
Linear Formula CuSO4
Fieser 01,164; 02,89; 05,162; 06,141; 08,125; 10,107; 16,283
Merck Index 15,2643
Solubility Information Solubility in water: 203g/L (20°C).
Formula Weight 159.6
Percent Purity ≥98%
Grade Analytical
Loss on Drying 1% (300°C) max.
Packaging Glass bottle
Chemical Name or Material Copper(II) sulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cupric sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.