missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10605492

Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10605492
500 g
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
5 kg
This item is not returnable. View return policy

Product Code. 10605492

Brand: Fisher Chemical C/8560/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number 149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110°C
Color Blue
pH 3.5 to 4.5
Physical Form Solid
Quantity 500 g
Formula Weight 249.68g/mol
Packaging Plastic powder jar
Vapor Pressure 7.3mmHg 25
compliance-icons compliance-icons
Product Identifier
  • COPPER (II) SULFATE PENTAHYDRATE
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate (1:1:5), Cupric sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) Sulfate Pentahydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.