missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Copper Oxide

Velp Scientifica™ Copper Oxide for use with Dumas Nitrogen Analyzer NDA 701 and 702.

Brand:  Velp Scientifica™ A00000157


Product Code. 15349574

  • 17.55 € / 50g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with NDA 701 and 702
Specifications

Specifications

Copper Oxide
50 g
Dumas Nitrogen Analyzer
0.05 kg
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • COPPER OXIDE, 50G

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : copper(II) oxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers