missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Appearance | Copper colored powder |
Assay Percent Range | 99.5% (Metals basis) |
Quantity | 500 g |
Mesh Size | -40+100 Mesh |
Odor | Odorless |
Form | Powder |
Chemical Name or Material | Copper |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S18G039 | Certificate of Analysis | 10/01/2023 | 000908.A3,000908.36 |
K08K29 | Certificate of Analysis | 08/12/2021 | 000908.A3,000908.36 |
F26K17 | Certificate of Analysis | 08/12/2021 | 000908.A3,000908.36 |
T24E064 | Certificate of Analysis | 08/12/2021 | 000908.A3,000908.36 |
K29S008 | Certificate of Analysis | 08/12/2021 | 000908.A3,000908.36 |
Safety and Handling


Product Identifier
- Copper powder
- Danger
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H411-Toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Copper powder, -40+100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction