Learn More
Copper Silicon bar, 3.8x3.8x30cm, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar 043767.KG
Product Code. 15461927
  • 197.00 € / Each
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Copper Silicon bar
Cu:Si; 97:3 wt%
1 Pc.
≈3.7kg/bar
3.8x3.8x30cm
MFCD02684526
Everdur™ Silicon Bronze
Bar