missing translation for 'onlineSavingsMsg'
Learn More

Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™

Product Code. 10743173
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Copper Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3251-23-8
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.554
MDL Number 10967
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
Synonym copper dinitrate, cupric nitrate, copper ii nitrate, copper nitrate, cupric dinitrate, claycop, copper 2+ nitrate, caswell no. 246, nitric acid, copper 2+ salt, unii-9tc879s2zv
PubChem CID 18616
ChEBI CHEBI:78036
IUPAC Name copper;dinitrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Cu+2]

Specifications

Color Colorless
pH 1
Quantity 100 mL
Packaging HDPE plastic bottle
Form Liquid
compliance-icons
Product Identifier
  • Copper solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.