missing translation for 'onlineSavingsMsg'
Learn More

Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 195931000


 View more versions of this product

Product Code. 10666022

  • 19.10 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Copper standard solution
1 mg/ml Cu in 2% HNO3
Plastic bottle
Blue
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
63.55
8.92
Solution
997 to 1003 μg/mL of Cu(2+) in 2% HNO3 (20°C)
7440-50-8, 7697-37-2, 7732-18-5
100 mL
Cu
16,95; 16,181
[Cu]
63.55
CHEBI:30052
AAS
Soluble in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Copper atomic absorption standard solution

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only