missing translation for 'onlineSavingsMsg'
Learn More

Copper standard solution, for AAS, 1 mg/ml Cu in 2% HNO3, Thermo Scientific Chemicals

19.10 € - 58.09 €

Specifications

Chemical Name or Material Copper standard solution
Concentration 997 to 1003 μg/mL of Cu(2+) in 2% HNO3 (20°C)
Name Note 1 mg/ml Cu in 2% HNO3
CAS 7440-50-8, 7697-37-2, 7732-18-5
Packaging Plastic bottle
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10666022
View Documents
Thermo Scientific Acros
195931000
100 mL Plastic bottle
19.10 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10419900
View Documents
Thermo Scientific Acros
195935000
500 mL Plastic bottle
58.09 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Copper standard solution
1 mg/ml Cu in 2% HNO3
Plastic bottle
Cu
16,95; 16,181
[Cu]
63.55
CHEBI:30052
AAS
Soluble in water
997 to 1003 μg/mL of Cu(2+) in 2% HNO3 (20°C)
7440-50-8, 7697-37-2, 7732-18-5
Blue
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
23978
63.55
8.92
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May be corrosive to metals.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep only in original container.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • Copper atomic absorption standard solution

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only