Learn More
JULABO CORIO CP Refrigerated/Heating Circulator
Suitable for applications with a temperature range up to +200°C. The enhanced pump performance ensures they are suitable for easy temperature control tasks in combination with external applications.
Brand:  JULABO 9013714.S1
 View more versions of this product
Product Code. 17724179
  • 7125.00 € / Each
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Models for internal and external applications
  • Bright, white, easy to read display
  • Very quiet
  • Easy pump change-over between internal and external circulation
  • External pump connections
  • Powerful and infinitely adjustable pressure pump
  • Bath lid and drain tap included
  • Removable ventilation grid
  • Refrigeration unit without side vents
  • Unique early warning system for low liquid level
  • Class III (FL) according to DIN 12876-1
Specifications
Specifications
3 to 4 L
USB, RS232
8 to 27 L/min.
-30°C to +200°C
CP-450F
15.7 x 9.1 x 25.6 in.
CE
440 W
Centrifugal
Circulator
R449A
230 V