Learn More
Fisherbrand™ Cork Borer
Manufactured from brass and nickel-plated.
15.35 € - 29.10 €
Specifications
Product Type Cork Borer
Material Nickel-plated brass
View More Specs
Products 9
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
12900896
View Documents
Fisherbrand™
3404/5
11.25mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12990886
View Documents
Fisherbrand™
3404/5
10mm
16.45 €
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
12960896
View Documents
Fisherbrand™
3405/18
26.25mm
29.10 €
Each
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
12910896
View Documents
Fisherbrand™
3404/7
12.5mm
16.15 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
12980886
View Documents
Fisherbrand™
3404/4
8.75mm
16.15 €
Each
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
12970886
View Documents
Fisherbrand™
3404/3
7.5mm
16.15 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
12902580
View Documents
Fisherbrand™
3404/2
6.25mm
16.45 €
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
13451900
View Documents
Fisherbrand™
3405/11
17.50mm
29.10 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
15147472
View Documents
Fisherbrand™
3404/8
13.75mm
16.15 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
Description
Description
  • Comes complete with push rod for clearing borers
  • All sizes are 100mm in length
  • Tube wall thickness of 0.45mm
Specifications
Specifications
Cork Borer
Nickel-plated brass
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Cork Borer