missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 0.25% Trypsin, 0.1% EDTA in HBSS w/o Calcium, Magnesium and Sodium Bicarbonate

Used to enzymatically release adherent cells from tissue culture plates for passaging.
Brand: Corning™ 25-053-CI
128.02 EUR valid until 2025-12-31
Use promo code "24335" to get your promotional price.
This item is not returnable.
View return policy
Description
- EDTA enhances efficacy of trypsin
- Manufactured under industry-leading standards for low endotoxin content and superior results
- Certificate of Analysis available
- Enzymatic dissociation reagents tested for porcine parvovirus

Specifications
-20°C | |
Cell Culture Dissociation Reagent | |
No Calcium, No Magnesium, No Sodium Bicarbonate | |
6 x 100 mL |
Liquid | |
Sterile | |
7 to 7.6 |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
27524003 | Certificate of Quality | 30/04/2025 | 25-053-CI |
17824005 | Certificate of Quality | 22/01/2025 | 25-053-CI |
14524002 | Certificate of Quality | 26/11/2024 | 25-053-CI |
07524002 | Certificate of Quality | 25/09/2024 | 25-053-CI |
16023003 | Certificate of Analysis | 19/07/2023 | 25-053-CI |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 0.25% Trypsin, 0.1% EDTA in HBSS w/o Calcium, Magnesium and Sodium Bicarbonate > 0.25% Trypsin, 0.1% EDTA in HBSS; w/o Ca, Mg, sodium bicarbonate; 6 x 100mL
Spot an opportunity for improvement?Share a Content Correction