missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Universal CellSTACK™ Caps
BIOPHARMAGREENER_CHOICE

For filling, venting, and manipulation

Brand:  Corning™ 3334

Product Code. 10211303

  • 313.00 € / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 33 mm threaded cap
  • Solid cap is used in closed systems or applications where venting is not necessary
  • Sterilized by gamma irradiation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Orange
Orange
Male MPC Polycarbonate, 0.25 in. I.D. coupling and female MPC Polycarbonate end cap
1/Pk.
Single-use
Screw Cap
HDPE
HDPE
4/Cs.
Cap, Filling
Sterile
33 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers