missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Shop All Corning Products

Click to view available options
Color:
Black
White
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Surface Treatment:
High Binding
Non-binding
Non-treated
Unit Size:
Pack of 50
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Specifications
| Format | Standard |
| Description | (NBS) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. |
| Array | 16 x 24 |
| Binding Type | Non Binding |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-binding |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction