missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand: Corning™ 3574
This item is not returnable.
View return policy
Description
- Minimal well to well crosstalk
- White microplates enhance luminecent signals and have low background luminescence
- Black mircoplates have low background fluorescence and minimize light scattering
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Medical-grade polystyrene
- Well volume: 112μL
- Working volume: 20μL to 80μL
- Surface area/well: 0.06cm2
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Plate dimensions meet proposed industry standards
Specifications
Standard | |
(NBS) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. | |
White | |
384 | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. |
Without Lid | |
16 x 24 | |
Polystyrene | |
20 to 80 μL | |
Non-binding | |
Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction