missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE

Product Code. 15314322 Shop All Corning Products
Click to view available options
Color:
Black
White
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Surface Treatment:
High Binding
Non-binding
Non-treated
Unit Size:
Pack of 50

Product Code. 15314322

Brand: Corning™ 4514

Only null left
Add to basket
Add to basket

Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.

 

  • Minimal well to well crosstalk
  • White microplates enhance luminecent signals and have low background luminescence
  • Black mircoplates have low background fluorescence and minimize light scattering
  • Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
  • Medical-grade polystyrene
  • Well volume: 112μL
  • Working volume: 20μL to 80μL
  • Surface area/well: 0.06cm2
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Plate dimensions meet proposed industry standards

 


TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Description Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
Array 16 x 24
Color Black
Product Type 384-Well Low Volume Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 2 to 25 μL
Sterility Non-sterile
Surface Treatment Non-binding
Well Shape Round Bottom (U-well)
For Use With (Application) Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.