Learn More
Corning™ 384-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE
Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Solid Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Brand:  Corning™ 3572
397.24 EUR valid until 2024-12-31
Use promo code "21693" to get your promotional price.
Product Code. 10109632
  • 461.63 € / Pack of 50
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
16 x 24
Polystyrene
20 to 80 μL
Non-treated
Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
10/Pk.
Without Lid
White
384
Non-sterile
Flat Bottom (F-well)
50/Cs.
50/Cs.