missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 384-Well Low-Volume Solid Microplates

Product Code. 10167431 Shop All Corning Products
Click to view available options
Color:
Black
White
Surface Treatment:
Non-binding
Non-treated
Tissue Culture
Volume (Metric) Working:
2 to 20 μL
2 to 25 μL
5 to 40 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 10167431

Brand: Corning™ 3677

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Same low reagent volumes as 1536-well plates without the need for new instrumentation

Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.

  • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
  • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
  • Choose black plates for fluorescence applications or white for luminescence applications
High-Binding Surface Plates
  • Ideal for immunoassays
  • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Working volume: 2 to 20μL

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Array 16 x 24
Color Black
Material Polystyrene
Volume (Metric) Well 35 μL
No. of Wells 384
Volume (Metric) Working 2 to 25 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
Well Color Black
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.