Learn More
Corning™ 384-Well Low-Volume Solid Microplates
GREENER_CHOICE
Same low reagent volumes as 1536-well plates without the need for new instrumentation
Brand:  Corning™ 3825
 View more versions of this product
Product Code. 10679029
  • 553.47 € / Pack of 50
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polystyrene plates feature a unique square-to-round well design with a conical-shaped bottom that provides easier access and imaging for superior assay performance.
  • Bottom reduces fluid and air entrapment, greater signal-to-noise ratio and reduced FP signal (mP) when working with very low volumes
  • Each well has a raised rim to reduce cross-contamination during assay and liquid-handling process
  • Choose black plates for fluorescence applications or white for luminescence applications
High-Binding Surface Plates
  • Ideal for immunoassays
  • Can bind medium (>10kDa) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Working volume: 2 to 20μL
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
16 x 24
Polystyrene
5 to 40 μL
Non-treated
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
White
Standard
White
384
Non-sterile
Flat Bottom (F-well)
50/Cs.